Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl (3-fluorobenzoyl)acetate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl (3-fluorobenzoyl)acetate
CAS 33166-77-7
Formula Weight 210.2
MDL Number MFCD03424809
Synonym 3-Fluorobenzoylacetic acid ethyl ester, 98%
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6131503

  SDS
alfa aesar™
H6131503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6131506

  SDS
alfa aesar™
H6131506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61315MD

  SDS
alfa aesar™
H61315MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.2
Boiling Point 262° to 263°C
MDL Number MFCD03424809
CAS 33166-77-7
Density 1.166g/mL
Chemical Name or Material Ethyl (3-fluorobenzoyl)acetate
Refractive Index 1.5170
Assay 98%
Synonym 3-Fluorobenzoylacetic acid ethyl ester, 98%
Color Colorless to orange
Quantity 1g
Physical Form Liquid
TSCA TSCA