missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3-fluorobenzoate, 98+%, Thermo Scientific Chemicals
$61.96 - $160.32
Chemical Identifiers
CAS | 451-02-5 |
---|---|
Molecular Formula | C9H9FO2 |
Molecular Weight (g/mol) | 168.167 |
MDL Number | MFCD00000336 |
InChI Key | SMMIKBXLEWTSJD-UHFFFAOYSA-N |
Synonym | ethyl m-fluorobenzoate, benzoic acid, 3-fluoro-, ethyl ester, 3-fluorobenzoic acid ethyl ester, 3-fluoro-benzoic acid ethyl ester, benzoic acid, m-fluoro-, ethyl ester, pubchem3470, 3-fluorobenzoic acid ethyl, acmc-209k0o, ethyl3-fluorobenzoate, 3-fluoro-benzoicaciethylester |
PubChem CID | 67975 |
IUPAC Name | ethyl 3-fluorobenzoate |
SMILES | CCOC(=O)C1=CC(=CC=C1)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0798409
|
Thermo Scientific Chemicals
L0798409 |
10 g |
Each for $61.96
|
|
|||||
AAL0798418
|
Thermo Scientific Chemicals
L0798418 |
50 g |
Each for $160.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
451-02-5 | |
168.167 | |
SMMIKBXLEWTSJD-UHFFFAOYSA-N | |
67975 | |
CCOC(=O)C1=CC(=CC=C1)F |
C9H9FO2 | |
MFCD00000336 | |
ethyl m-fluorobenzoate, benzoic acid, 3-fluoro-, ethyl ester, 3-fluorobenzoic acid ethyl ester, 3-fluoro-benzoic acid ethyl ester, benzoic acid, m-fluoro-, ethyl ester, pubchem3470, 3-fluorobenzoic acid ethyl, acmc-209k0o, ethyl3-fluorobenzoate, 3-fluoro-benzoicaciethylester | |
ethyl 3-fluorobenzoate |
Specifications
451-02-5 | |
208°C | |
Recognizable | |
1.485 | |
10 g | |
ethyl m-fluorobenzoate, benzoic acid, 3-fluoro-, ethyl ester, 3-fluorobenzoic acid ethyl ester, 3-fluoro-benzoic acid ethyl ester, benzoic acid, m-fluoro-, ethyl ester, pubchem3470, 3-fluorobenzoic acid ethyl, acmc-209k0o, ethyl3-fluorobenzoate, 3-fluoro-benzoicaciethylester | |
CCOC(=O)C1=CC(=CC=C1)F | |
168.167 | |
168.17 | |
Ethyl 3-fluorobenzoate |
1.136 | |
82°C (179°F) | |
C9H9FO2 | |
MFCD00000336 | |
2045876 | |
SMMIKBXLEWTSJD-UHFFFAOYSA-N | |
ethyl 3-fluorobenzoate | |
67975 | |
≥98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 207-191-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only