Please login to your online account to display your discounted pricing

Ethyl 3-exo-aminobicyclo[2.2.1]heptane-2-exo-carboxylate hydrochloride, 98%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  265722500

 View more versions of this product

Catalog No. AC265722500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 219.71
Quantity 250mg
Color White
Chemical Name or Material Ethyl 3-exo-aminobicyclo[2.2.1]heptane-2-exo-carboxylate hydrochloride
Packaging Glass bottle
Name Note 98%
CAS 95630-75-4
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C10H17NO2·HCl
MDL Number MFCD00144273

Specifications

Chemical Name or Material Ethyl 3-exo-aminobicyclo[2.2.1]heptane-2-exo-carboxylate hydrochloride
Name Note 98%
CAS 95630-75-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H17NO2·HCl
Formula Weight 219.71
MDL Number MFCD00144273