Please login to your online account to display your discounted pricing

Ethyl 3-exo-aminobicyclo[2.2.1]heptane-2-exo-carboxylate hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 3-exo-aminobicyclo[2.2.1]heptane-2-exo-carboxylate hydrochloride
Name Note 98%
CAS 95630-75-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H17NO2·HCl
Formula Weight 219.71
MDL Number MFCD00144273
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC265722500

  SDS
thermo fisher scientific
265722500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 219.71
Quantity 250mg
Color White
Chemical Name or Material Ethyl 3-exo-aminobicyclo[2.2.1]heptane-2-exo-carboxylate hydrochloride
Packaging Glass bottle
Name Note 98%
CAS 95630-75-4
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C10H17NO2·HCl
MDL Number MFCD00144273