missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 265732500

 View more versions of this product

Catalog No. AC265732500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride
Glass bottle
129°C to 132°C
99%
98.5% min. (Argentometry)
MFCD00144272
Solubility in water: soluble. Other solubilities: soluble in ethanol
CCOC(=O)C1C2CC(C1N)C=C2.Cl
217.69
217.69
99%
Authentic
White to Beige
250mg
104770-18-5
C10H15NO2·HCl
ethyl 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-carboxylate hydrochloride, ethyl 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-exo-carboxylate hydrochloride
PBLUKJJZYDTALO-WZSOELRKSA-N
ethyl (1S,2S,3R,4R)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylate;hydrochloride
71528337
Fine Crystalline Powder

Chemical Identifiers

104770-18-5
217.69
PBLUKJJZYDTALO-WZSOELRKSA-N
71528337
CCOC(=O)C1C2CC(C1N)C=C2.Cl
C10H15NO2·HCl
MFCD00144272
ethyl 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-carboxylate hydrochloride, ethyl 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-exo-carboxylate hydrochloride
ethyl (1S,2S,3R,4R)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylate;hydrochloride
SDS Documents