Please login to your online account to display your discounted pricing

Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride
Name Note 99%
CAS 104770-18-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H15NO2·HCl
Formula Weight 217.69
MDL Number MFCD00144272
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC265730010

  SDS
thermo fisher scientific
265730010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC265732500

  SDS
thermo fisher scientific
265732500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 217.69
Quantity 1g
Color White to light beige
Chemical Name or Material Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H15NO2·HCl
MDL Number MFCD00144272
CAS 104770-18-5
Infrared Spectrum Authentic