missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Ethoxypropionate, TCI America™

Manufacturer:  TCI America E0319500ML

 View more versions of this product

Catalog No. E0319500ML



Specifications

Specifications

Ethyl 3-Ethoxypropionate
Colorless
763-69-9
MFCD00051356
ethyl 3-ethoxypropionate, 3-ethoxypropionic acid ethyl ester, propanoic acid, 3-ethoxy-, ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl beta-ethoxypropionate, unii-ec38rsj79j, propionic acid, 3-ethoxy-, ethyl ester, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove
CCOCCC(=O)OCC
146.186
146.19
≥99.0% (GC)
170°C
500mL
C7H14O3
3272
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
ethyl 3-ethoxypropanoate
12989
Clear Liquid at 20°C

Chemical Identifiers

763-69-9
146.186
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
12989
CCOCCC(=O)OCC
C7H14O3
MFCD00051356
ethyl 3-ethoxypropionate, 3-ethoxypropionic acid ethyl ester, propanoic acid, 3-ethoxy-, ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl beta-ethoxypropionate, unii-ec38rsj79j, propionic acid, 3-ethoxy-, ethyl ester, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove
ethyl 3-ethoxypropanoate
Safety and Handling

Safety and Handling

RTECSNumber : UF3325000

TSCA : Yes

Documents