Learn More
Ethyl 3-ethoxypropionate, 99+%, pure, stabilized, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 269490010
Specifications
Ethyl 3-ethoxypropionate | |
59°C | |
Glass bottle | |
0.949 | |
1.3 mPa.s (20°C) | |
170°C | |
-75°C | |
99+% | |
99% min. (GC) | |
C2H5OC(O)CH2CH2OC2H5 | |
3-ethoxypropionic acid ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl 3-ethoxypropionate, ethyl beta-ethoxypropionate, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove, propanoic acid, 3-ethoxy-, ethyl ester, propionic acid, 3-ethoxy-, ethyl ester, unii-ec38rsj79j | |
BHXIWUJLHYHGSJ-UHFFFAOYSA-N | |
ethyl 3-ethoxypropanoate | |
12989 | |
Liquid | |
Pure |
0.9490g/mL | |
Authentic | |
1.4060 to 1.408 | |
50 to 100ppm BHT | |
0.05% max. | |
Undesignated | |
1L | |
763-69-9 | |
C7H14O3 | |
MFCD00051356 | |
Solubility in water: 1.6% (20 c) | |
CCOCCC(=O)OCC | |
146.19 | |
146.19 | |
99+% |
Chemical Identifiers
763-69-9 | |
146.19 | |
BHXIWUJLHYHGSJ-UHFFFAOYSA-N | |
12989 | |
CCOCCC(=O)OCC |
C7H14O3 | |
MFCD00051356 | |
3-ethoxypropionic acid ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl 3-ethoxypropionate, ethyl beta-ethoxypropionate, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove, propanoic acid, 3-ethoxy-, ethyl ester, propionic acid, 3-ethoxy-, ethyl ester, unii-ec38rsj79j | |
ethyl 3-ethoxypropanoate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 212-112-9
RTECSNumber : UF3325000
TSCA : TSCA