missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-ethoxypropionate, 99+%, pure, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC269490100

 View more versions of this product

Catalog No. AC269490100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl 3-ethoxypropionate
59°C
Plastic drum
0.949
1.3 mPa.s (20°C)
170°C
-75°C
99+%
99% min. (GC)
C2H5OC(O)CH2CH2OC2H5
3-ethoxypropionic acid ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl 3-ethoxypropionate, ethyl beta-ethoxypropionate, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove, propanoic acid, 3-ethoxy-, ethyl ester, propionic acid, 3-ethoxy-, ethyl ester, unii-ec38rsj79j
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
ethyl 3-ethoxypropanoate
12989
Liquid
Pure
0.9490g/mL
Authentic
1.4060 to 1.408
50 to 100ppm BHT
0.05% max.
Undesignated
10L
763-69-9
C7H14O3
MFCD00051356
Solubility in water: 1.6% (20 c)
CCOCCC(=O)OCC
146.19
146.19
99+%

Chemical Identifiers

763-69-9
146.19
BHXIWUJLHYHGSJ-UHFFFAOYSA-N
12989
CCOCCC(=O)OCC
C7H14O3
MFCD00051356
3-ethoxypropionic acid ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl 3-ethoxypropionate, ethyl beta-ethoxypropionate, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove, propanoic acid, 3-ethoxy-, ethyl ester, propionic acid, 3-ethoxy-, ethyl ester, unii-ec38rsj79j
ethyl 3-ethoxypropanoate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-112-9

RTECSNumber : UF3325000

TSCA : TSCA

SDS Documents
Product Certifications