Learn More
Ethyl 3-ethoxyacrylate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5918718
Description
Used in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the preparation of N-[3-(3-Cyanopyrazolo[1,5-a]pyrimidin-5-yl)phenyl]-N-ethylacetamide.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Ethyl 3-ethoxyacrylate | |
Colorless | |
195°C to 196°C | |
C7H12O3 | |
CH3CH2OCH=CHCOOCH2CH3 | |
50 g | |
1721612 | |
Slightly soluble in water. | |
CCOC=CC(=O)OCC | |
144.17 | |
144.17 | |
Liquid |
1001-26-9 | |
0.998 g/mL | |
76°C (169°F) | |
1.4465 | |
MFCD00009863 | |
UN3334 | |
ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate | |
ITQFPVUDTFABDH-AATRIKPKSA-N | |
ethyl (E)-3-ethoxyprop-2-enoate | |
5366269 | |
98% |
Chemical Identifiers
1001-26-9 | |
144.17 | |
ITQFPVUDTFABDH-AATRIKPKSA-N | |
5366269 | |
CCOC=CC(=O)OCC |
C7H12O3 | |
MFCD00009863 | |
ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate | |
ethyl (E)-3-ethoxyprop-2-enoate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H227-H315-H319
DOTInformation : Hazard Class: 9; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only