missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3-ethoxy-3-iminopropionate hydrochloride, 85+%, ACROS Organics™
Chemical Identifiers
CAS | 2318-25-4 |
---|---|
Molecular Formula | C7H13NO3·HCl |
Molecular Weight (g/mol) | 195.64 |
MDL Number | MFCD00051405 |
InChI Key | HYMXUYQKXCHWDC-UHFFFAOYSA-N |
Synonym | 3-ethoxy-3-iminopropanoic acid ethyl ester hydrochloride, diethyl malonmonoimidate hydrochloride, ethyl 3-ethoxy-3-iminopropanoate hydrochloride, ethyl 3-ethoxy-3-iminopropionate hcl, ethyl 3-ethoxy-3-iminopropionate hydrochloride, ethyl beta-ethoxy-beta-iminopropionate hydrochloride, ethyl carbethoxyacetimidate hydrochloride, ethyl ethoxycarbonylacetimidate hydrochloride, ethyl3-ethoxy-3-iminopropionate hydrochloride, propanoic acid, 3-ethoxy-3-imino-, ethyl ester, hydrochloride |
PubChem CID | 75345 |
IUPAC Name | ethyl 3-ethoxy-3-iminopropanoate;hydrochloride |
SMILES | CCOC(=N)CC(=O)OCC.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC399500050
|
Thermo Fisher Scientific
AC399500050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC399500250
|
Thermo Fisher Scientific
AC399500250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC399501000
|
Thermo Fisher Scientific
AC399501000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
2318-25-4 | |
195.64 | |
HYMXUYQKXCHWDC-UHFFFAOYSA-N | |
75345 | |
CCOC(=N)CC(=O)OCC.Cl |
C7H13NO3·HCl | |
MFCD00051405 | |
3-ethoxy-3-iminopropanoic acid ethyl ester hydrochloride, diethyl malonmonoimidate hydrochloride, ethyl 3-ethoxy-3-iminopropanoate hydrochloride, ethyl 3-ethoxy-3-iminopropionate hcl, ethyl 3-ethoxy-3-iminopropionate hydrochloride, ethyl beta-ethoxy-beta-iminopropionate hydrochloride, ethyl carbethoxyacetimidate hydrochloride, ethyl ethoxycarbonylacetimidate hydrochloride, ethyl3-ethoxy-3-iminopropionate hydrochloride, propanoic acid, 3-ethoxy-3-imino-, ethyl ester, hydrochloride | |
ethyl 3-ethoxy-3-iminopropanoate;hydrochloride |
Specifications
Glass bottle | |
5g | |
85% min. (Argentometry) | |
CH3CH2OC(=NH)CH2COOCH2CH3·HCL | |
3-ethoxy-3-iminopropanoic acid ethyl ester hydrochloride, diethyl malonmonoimidate hydrochloride, ethyl 3-ethoxy-3-iminopropanoate hydrochloride, ethyl 3-ethoxy-3-iminopropionate hcl, ethyl 3-ethoxy-3-iminopropionate hydrochloride, ethyl beta-ethoxy-beta-iminopropionate hydrochloride, ethyl carbethoxyacetimidate hydrochloride, ethyl ethoxycarbonylacetimidate hydrochloride, ethyl3-ethoxy-3-iminopropionate hydrochloride, propanoic acid, 3-ethoxy-3-imino-, ethyl ester, hydrochloride | |
HYMXUYQKXCHWDC-UHFFFAOYSA-N | |
ethyl 3-ethoxy-3-iminopropanoate;hydrochloride | |
75345 | |
85+% |
108°C to 110°C | |
2318-25-4 | |
C7H13NO3·HCl | |
MFCD00051405 | |
Solubility in water: decomposes | |
CCOC(=N)CC(=O)OCC.Cl | |
195.64 | |
195.64 | |
Ethyl 3-ethoxy-3-iminopropionate hydrochloride |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 219-028-1