missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-ethoxy-3-iminopropionate hydrochloride, 85+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC399500250

 View more versions of this product

Catalog No. AC399500250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl 3-ethoxy-3-iminopropionate hydrochloride
108°C to 110°C
2318-25-4
C7H13NO3·HCl
MFCD00051405
Solubility in water: decomposes
CCOC(=N)CC(=O)OCC.Cl
195.64
195.64
Glass bottle
25g
85% min. (Argentometry)
CH3CH2OC(=NH)CH2COOCH2CH3·HCL
3-ethoxy-3-iminopropanoic acid ethyl ester hydrochloride, diethyl malonmonoimidate hydrochloride, ethyl 3-ethoxy-3-iminopropanoate hydrochloride, ethyl 3-ethoxy-3-iminopropionate hcl, ethyl 3-ethoxy-3-iminopropionate hydrochloride, ethyl beta-ethoxy-beta-iminopropionate hydrochloride, ethyl carbethoxyacetimidate hydrochloride, ethyl ethoxycarbonylacetimidate hydrochloride, ethyl3-ethoxy-3-iminopropionate hydrochloride, propanoic acid, 3-ethoxy-3-imino-, ethyl ester, hydrochloride
HYMXUYQKXCHWDC-UHFFFAOYSA-N
ethyl 3-ethoxy-3-iminopropanoate;hydrochloride
75345
85+%

Chemical Identifiers

2318-25-4
195.64
HYMXUYQKXCHWDC-UHFFFAOYSA-N
75345
CCOC(=N)CC(=O)OCC.Cl
C7H13NO3·HCl
MFCD00051405
3-ethoxy-3-iminopropanoic acid ethyl ester hydrochloride, diethyl malonmonoimidate hydrochloride, ethyl 3-ethoxy-3-iminopropanoate hydrochloride, ethyl 3-ethoxy-3-iminopropionate hcl, ethyl 3-ethoxy-3-iminopropionate hydrochloride, ethyl beta-ethoxy-beta-iminopropionate hydrochloride, ethyl carbethoxyacetimidate hydrochloride, ethyl ethoxycarbonylacetimidate hydrochloride, ethyl3-ethoxy-3-iminopropionate hydrochloride, propanoic acid, 3-ethoxy-3-imino-, ethyl ester, hydrochloride
ethyl 3-ethoxy-3-iminopropanoate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 219-028-1

SDS Documents