Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-ethoxy-2-butenoate, 97%

$145.70 - $145.70

Chemical Identifiers

Chemical Name or Material Ethyl 3-ethoxy-2-butenoate
CAS 998-91-4
Molecular Formula C8H14O3
Formula Weight 158.2
MDL Number MFCD00026932
Beilstein 1757006
Synonym 3-Ethoxy-2-butenoic acid ethyl ester; Ethyl 3-ethoxycrotonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1109714

  SDS
alfa aesar™
A1109714
25g Each for $145.70
Description & Specifications

Specifications

Formula Weight 158.2
Assay 97%
Beilstein 1757006
Molecular Formula C8H14O3
Melting Point 28° to 32°C
CAS 998-91-4
Boiling Point 86° to 88°C (14mmHg)
Quantity 25g
Synonym 3-Ethoxy-2-butenoic acid ethyl ester; Ethyl 3-ethoxycrotonate
MDL Number MFCD00026932
Chemical Name or Material Ethyl 3-ethoxy-2-butenoate

Applications
Ethyl 3-ethoxy-2-butenoate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.