Please login to your online account to display your discounted pricing

Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride, 98%, Acros Organics


Quantity 250mg

Manufacturer: thermo fisher scientific  265712500

 View more versions of this product

Catalog No. AC265712500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 217.69
Quantity 250mg
Color White to beige
Infrared Spectrum Authentic
Name Note 98%
Molecular Formula C10H15NO2·HCl
Chemical Name or Material Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride
MDL Number MFCD00144270
Assay Percent Range 98%
CAS 95630-74-3

Specifications

Chemical Name or Material Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride
Name Note 98%
CAS 95630-74-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H15NO2·HCl
Formula Weight 217.69
MDL Number MFCD00144270