Please login to your online account to display your discounted pricing

Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride, 98%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  265710010

 View more versions of this product

Catalog No. AC265710010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 217.69
Physical Form Crystalline Powder
Assay ≥97.5%
Quantity 1g
Color White to beige
Infrared Spectrum Authentic
Molecular Formula C10H15NO2·HCl
MDL Number MFCD00144270
Name Note 98%
Chemical Name or Material Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride
CAS 95630-74-3
Assay Percent Range 98%

Specifications

Chemical Name or Material Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride
Name Note 98%
CAS 95630-74-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H15NO2·HCl
Formula Weight 217.69
MDL Number MFCD00144270