Please login to your online account to display your discounted pricing

Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride
Name Note 98%
CAS 95630-74-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H15NO2·HCl
Formula Weight 217.69
MDL Number MFCD00144270
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC265710010

  SDS
thermo fisher scientific
265710010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC265712500

  SDS
thermo fisher scientific
265712500
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 217.69
Physical Form Crystalline Powder
Assay ≥97.5%
Quantity 1g
Color White to beige
Infrared Spectrum Authentic
Molecular Formula C10H15NO2·HCl
MDL Number MFCD00144270
Name Note 98%
Chemical Name or Material Ethyl 3-endo-aminobicyclo[2.2.1]hept-5-ene-2-endo-carboxylate hydrochloride
CAS 95630-74-3
Assay Percent Range 98%