missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-cyano-3-phenylpyruvate, 97%, Thermo Scientific Chemicals

$52.74 - $184.03

Chemical Identifiers

CAS 6362-63-6
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.22
MDL Number MFCD00019801
InChI Key YWNRATDDUSMBPR-UHFFFAOYNA-N
Synonym ethyl 3-cyano-3-phenylpyruvate, ethyl phenylcyanopyruvate, cyanophenylpyruvic acid ethyl ester, ethyl cyanophenylpyruvate, ethyl alpha-cyanophenylpyruvate, pyruvic acid, cyanophenyl-, ethyl ester, ethyl .alpha.-cyanophenylpyruvate, wln: ncr x1v1uno2, 4-10-00-03440 beilstein handbook reference, benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl ester
PubChem CID 22845
IUPAC Name ethyl 3-cyano-2-oxo-3-phenylpropanoate
SMILES CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2695106
View Documents Promotion Details
Thermo Scientific Chemicals
H2695106
5 g
Each for $52.74
 
AAH2695114
View Documents Promotion Details
Thermo Scientific Chemicals
H2695114
25 g
Each for $184.03
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6362-63-6
217.22
YWNRATDDUSMBPR-UHFFFAOYNA-N
22845
CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1
C12H11NO3
MFCD00019801
ethyl 3-cyano-3-phenylpyruvate, ethyl phenylcyanopyruvate, cyanophenylpyruvic acid ethyl ester, ethyl cyanophenylpyruvate, ethyl alpha-cyanophenylpyruvate, pyruvic acid, cyanophenyl-, ethyl ester, ethyl .alpha.-cyanophenylpyruvate, wln: ncr x1v1uno2, 4-10-00-03440 beilstein handbook reference, benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl ester
ethyl 3-cyano-2-oxo-3-phenylpropanoate

Specifications

6362-63-6
C12H11NO3
5 g
YWNRATDDUSMBPR-UHFFFAOYNA-N
ethyl 3-cyano-2-oxo-3-phenylpropanoate
22845
97%
125°C to 129°C
MFCD00019801
ethyl 3-cyano-3-phenylpyruvate, ethyl phenylcyanopyruvate, cyanophenylpyruvic acid ethyl ester, ethyl cyanophenylpyruvate, ethyl alpha-cyanophenylpyruvate, pyruvic acid, cyanophenyl-, ethyl ester, ethyl .alpha.-cyanophenylpyruvate, wln: ncr x1v1uno2, 4-10-00-03440 beilstein handbook reference, benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl ester
CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1
217.22
217.23
Ethyl 3-cyano-3-phenylpyruvate
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 228-844-7

RTECSNumber : UZ0875000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only