Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-chloropropionate, 98%

$50.18 - $255.53

Chemical Identifiers

Chemical Name or Material Ethyl 3-chloropropionate
CAS 623-71-2
Molecular Formula ClCH2CH2CO2CH2CH3
Formula Weight 136.58
MDL Number MFCD00000989
Beilstein 385698
Synonym 3-Chloropropionic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2390714

 
alfa aesar™
B2390714
25g Each for $50.18

AAB2390722

 
alfa aesar™
B2390722
100g Each for $142.07

AAB2390730

 
alfa aesar™
B2390730
250g Each for $255.53
Description & Specifications

Specifications

Formula Weight 136.58
Chemical Name or Material Ethyl 3-chloropropionate
TSCA TSCA
Assay 98%
Molecular Formula ClCH2CH2CO2CH2CH3
MDL Number MFCD00000989
CAS 623-71-2
Beilstein 385698
Synonym 3-Chloropropionic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 162° to 164°C
Quantity 25g
Flash Point 61°C (142°F)

Applications
It can be used to produce (1-ethoxycyclopropyloxy)trimethylsilane by heating.

Solubility
Immiscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.