Please login to your online account to display your discounted pricing

Ethyl 3-chlorophenylacetate, +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 3-chlorophenylacetate
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C10H11ClO2
Linear Formula ClC6H4CH2(O)OCH2CH3
Formula Weight 198.65
MDL Number MFCD07787498
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388830250

  SDS
thermo fisher scientific
AC388830250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 198.65
Physical Form Liquid
Quantity 25g
Color Clear colorless
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Linear Formula ClC6H4CH2(O)OCH2CH3
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material Ethyl 3-chlorophenylacetate
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98+%
Molecular Formula C10H11ClO2
MDL Number MFCD07787498
CAS 14062-29-4
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.