Please login to your online account to display your discounted pricing

Ethyl 3-bromopropionate 99%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  AC409815000

 View more versions of this product

Catalog No. AC409815000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 181.03
Physical Form Liquid
Assay ≥98.5%
Quantity 500g
Color Clear colorless to pale yellow
Beilstein 02, 256
Merck Index 15, 1444
EINECS Number 208-724-0
TSCA TSCA
Density 1.4190g/mL
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00000251
Molecular Formula C5H9BrO2
Chemical Name or Material Ethyl 3-bromopropionate
Name Note 99%
Assay Percent Range 99%
Linear Formula CH2BrCH2COOC2H5
CAS 539-74-2

Specifications

Chemical Name or Material Ethyl 3-bromopropionate
Name Note 99%
CAS 539-74-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C5H9BrO2
Linear Formula CH2BrCH2COOC2H5
Formula Weight 181.03
MDL Number MFCD00000251
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents