Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-bromoindole-2-carboxylate, 97%

Chemical Identifiers

Chemical Name or Material Ethyl 3-bromoindole-2-carboxylate
CAS 91348-45-7
Formula Weight 268.11
MDL Number MFCD02071793
Beilstein 186192
Synonym 3-Bromo-indole-2-carboxylic acid ethyl ester, 97%
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6695103

 
alfa aesar™
H6695103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6695106

 
alfa aesar™
H6695106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 268.11
Chemical Name or Material Ethyl 3-bromoindole-2-carboxylate
Physical Form Solid
MDL Number MFCD02071793
CAS 91348-45-7
Beilstein 186192
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Quantity 1g
Synonym 3-Bromo-indole-2-carboxylic acid ethyl ester, 97%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.