Learn More
Ethyl 3-bromobenzoate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1025118
Description
Ethyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4'-methyl-biphenyl-3-carboxylic acid ethyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 3-bromobenzoate is used in the preparation of ethyl 3-ethynylbenzoate. It can react with p-tolyl-boronic acid, and obtain the 4′-methyl-biphenyl-3-carboxylic acid ethyl ester.
Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Ethyl 3-bromobenzoate | |
1.43 | |
>110°C (230°F) | |
1.543 | |
50 g | |
ethyl3-bromobenzoate, 3-bromobenzoic acid ethyl ester, benzoic acid, 3-bromo-, ethyl ester, ethyl-3-bromobenzoate, benzoic acid, m-bromo-, ethyl ester, m-bromo benzoic acid ethyl ester, m-bromobenzoic acid, ethyl ester, pubchem3941, acmc-209gbv, ethyl 3-bromobenzoate | |
CCOC(=O)C1=CC(=CC=C1)Br | |
229.073 | |
229.07 |
24398-88-7 | |
261°C | |
C9H9BrO2 | |
MFCD00013529 | |
1867121 | |
QAUASTLEZAPQTB-UHFFFAOYSA-N | |
ethyl 3-bromobenzoate | |
90488 | |
≥98% |
Chemical Identifiers
24398-88-7 | |
229.073 | |
QAUASTLEZAPQTB-UHFFFAOYSA-N | |
ethyl 3-bromobenzoate |
C9H9BrO2 | |
MFCD00013529 | |
90488 | |
CCOC(=O)C1=CC(=CC=C1)Br |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-223-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only