missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-bromobenzenesulfonate, 97%

Chemical Identifiers

CAS 1421262-56-7
Molecular Formula C8H9BrO3S
Molecular Weight (g/mol) 265.121
MDL Number MFCD22683135
InChI Key KVPGXLXCDWOMIC-UHFFFAOYSA-N
Synonym 3-Bromobenzenesulfonic acid ethyl ester
PubChem CID 87640011
IUPAC Name ethyl 3-bromobenzenesulfonate
SMILES CCOS(=O)(=O)C1=CC(=CC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60540MD View Documents Alfa Aesar™
H60540MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6054003 View Documents Alfa Aesar™
H6054003
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421262-56-7
265.121
KVPGXLXCDWOMIC-UHFFFAOYSA-N
87640011
CCOS(=O)(=O)C1=CC(=CC=C1)Br
C8H9BrO3S
MFCD22683135
3-Bromobenzenesulfonic acid ethyl ester
ethyl 3-bromobenzenesulfonate

Specifications

250mg
C8H9BrO3S
3-Bromobenzenesulfonic acid ethyl ester
CCOS(=O)(=O)C1=CC(=CC=C1)Br
265.121
265.12
Ethyl 3-bromobenzenesulfonate
1421262-56-7
MFCD22683135
KVPGXLXCDWOMIC-UHFFFAOYSA-N
ethyl 3-bromobenzenesulfonate
87640011
97%