missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Bromo-2-(bromomethyl)propionate, TCI America™

Manufacturer:  TCI America E079125G

 View more versions of this product

Catalog No. E079125G



Specifications

Specifications

Ethyl 3-Bromo-2-(bromomethyl)propionate
Undesignated
58539-11-0
MFCD00034154
GKSCTYSHDIGNGC-UHFFFAOYSA-N
ethyl 3-bromo-2-(bromomethyl)propanoate
310617
Clear Liquid at 20°C
86°C
25g
C6H10Br2O2
3-Bromo-2-(bromomethyl)propionic Acid Ethyl Ester
CCOC(=O)C(CBr)CBr
273.952
273.95
≥97.0% (GC)

Chemical Identifiers

58539-11-0
273.952
GKSCTYSHDIGNGC-UHFFFAOYSA-N
310617
CCOC(=O)C(CBr)CBr
C6H10Br2O2
MFCD00034154
3-Bromo-2-(bromomethyl)propionic Acid Ethyl Ester
ethyl 3-bromo-2-(bromomethyl)propanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents