missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Aminopyrazole-4-carboxylate, TCI America™

Manufacturer:  TCI America E076625G

 View more versions of this product

Catalog No. E076625G



Specifications

Specifications

Ethyl 3-Aminopyrazole-4-carboxylate
102°C
6994-25-8
MFCD00005238
YPXGHKWOJXQLQU-UHFFFAOYSA-N
ethyl 5-amino-1H-pyrazole-4-carboxylate
81472
Crystal-Powder at 20°C
Yellow
25g
C6H9N3O2
ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
CCOC(=O)C1=C(NN=C1)N
155.157
155.16
≥98.0% (GC,T)

Chemical Identifiers

6994-25-8
155.157
YPXGHKWOJXQLQU-UHFFFAOYSA-N
81472
CCOC(=O)C1=C(NN=C1)N
C6H9N3O2
MFCD00005238
ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
ethyl 5-amino-1H-pyrazole-4-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents