Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-aminobenzo[b]furan-2-carboxylate, 97%

$50.92 - $164.24

Chemical Identifiers

Chemical Name or Material Ethyl 3-aminobenzo[b]furan-2-carboxylate
CAS 39786-35-1
Molecular Formula C11H11NO3
Formula Weight 205.22
MDL Number MFCD00641330
Synonym 3-Aminobenzo[b]furan-2-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2712403

  SDS
alfa aesar™
H2712403
1g Each for $50.92

AAH2712406

  SDS
alfa aesar™
H2712406
5g Each for $164.24
Description & Specifications

Specifications

Formula Weight 205.22
Chemical Name or Material Ethyl 3-aminobenzo[b]furan-2-carboxylate
Assay 97%
Molecular Formula C11H11NO3
MDL Number MFCD00641330
CAS 39786-35-1
Synonym 3-Aminobenzo[b]furan-2-carboxylic acid ethyl ester
Melting Point 77° to 82°C
Quantity 1g