missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Amino-4-methylbenzoate, TCI America™

Manufacturer:  TCI America E102525G

 View more versions of this product

Catalog No. E102525G



Specifications

Specifications

Ethyl 3-Amino-4-methylbenzoate
White-Yellow
25g
C10H13NO2
3-Amino-4-methylbenzoic Acid Ethyl Ester, Ethyl 3-Amino-p-toluate, 3-Amino-p-toluic Acid Ethyl Ester
CCOC(=O)C1=CC(=C(C=C1)C)N
179.219
179.22
≥98.0% (GC,T)
105°C
51°C
41191-92-8
MFCD01926538
MCNBNDUVWQEKNZ-UHFFFAOYSA-N
ethyl 3-amino-4-methylbenzoate
12607966
Crystal-Powder at 20°C

Chemical Identifiers

41191-92-8
179.219
MCNBNDUVWQEKNZ-UHFFFAOYSA-N
12607966
CCOC(=O)C1=CC(=C(C=C1)C)N
C10H13NO2
MFCD01926538
3-Amino-4-methylbenzoic Acid Ethyl Ester, Ethyl 3-Amino-p-toluate, 3-Amino-p-toluic Acid Ethyl Ester
ethyl 3-amino-4-methylbenzoate
Safety and Handling

Safety and Handling

TSCA : No

Documents