missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-Acetoxyhexanoate, TCI America™

Manufacturer:  TCI America E101425G

 View more versions of this product

Catalog No. E101425G



Specifications

Specifications

Ethyl 3-Acetoxyhexanoate
Colorless
21188-61-4
MFCD00792508
UYBMIHNGXIOMAW-UHFFFAOYSA-N
ethyl 3-acetyloxyhexanoate
89464
Clear Liquid at 20°C
116°C
25g
C10H18O4
3-Acetoxyhexanoic Acid Ethyl Ester
CCCC(CC(=O)OCC)OC(=O)C
202.25
202.25
≥98.0% (GC)

Chemical Identifiers

21188-61-4
202.25
UYBMIHNGXIOMAW-UHFFFAOYSA-N
89464
CCCC(CC(=O)OCC)OC(=O)C
C10H18O4
MFCD00792508
3-Acetoxyhexanoic Acid Ethyl Ester
ethyl 3-acetyloxyhexanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents