Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,5-dinitrobenzoate, 98%

$37.36 - $37.36

Chemical Identifiers

Chemical Name or Material Ethyl 3,5-dinitrobenzoate
CAS 618-71-3
Molecular Formula C9H8N2O6
Formula Weight 240.17
MDL Number MFCD00007232
Beilstein 1995473
Synonym 3,5-Dinitrobenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2158218

  SDS
alfa aesar™
B2158218
50g Each for $37.36
Description & Specifications

Specifications

Formula Weight 240.17
Melting Point 91° to 92°C
MDL Number MFCD00007232
CAS 618-71-3
Assay 98%
Molecular Formula C9H8N2O6
Synonym 3,5-Dinitrobenzoic acid ethyl ester
Chemical Name or Material Ethyl 3,5-dinitrobenzoate
Beilstein 1995473
Quantity 50g