Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,5-dimethylphenylacetate, 98+%

$317.09 - $317.09

Chemical Identifiers

Chemical Name or Material Ethyl 3,5-dimethylphenylacetate
CAS 105337-18-6
Molecular Formula C12H16O2
Formula Weight 192.26
MDL Number MFCD00082645
Beilstein 2576084
Synonym 3,5-Dimethylphenylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1042309

 
alfa aesar™
L1042309
10g Each for $317.09
Description & Specifications

Specifications

Formula Weight 192.26
Chemical Name or Material Ethyl 3,5-dimethylphenylacetate
Assay ≥98%
MDL Number MFCD00082645
CAS 105337-18-6
Molecular Formula C12H16O2
Beilstein 2576084
Synonym 3,5-Dimethylphenylacetic acid ethyl ester
Boiling Point 138° to 139°C (15mmHg)
Quantity 10g