Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,5-dimethylbenzoate, 98+%

$406.82 - $406.82

Chemical Identifiers

Chemical Name or Material Ethyl 3,5-dimethylbenzoate
CAS 21239-29-2
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00015440
Beilstein 2088978
Synonym 3,5-Dimethylbenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0289014

  SDS
alfa aesar™
L0289014
25g Each for $406.82
Description & Specifications

Specifications

Formula Weight 178.23
CAS 21239-29-2
Chemical Name or Material Ethyl 3,5-dimethylbenzoate
Assay ≥98%
Molecular Formula C11H14O2
MDL Number MFCD00015440
Synonym 3,5-Dimethylbenzoic acid ethyl ester
Health Hazard 1 Warning
Odor Characteristic
Beilstein 2088978
Boiling Point 120°C (10mmHg)
Quantity 25g
Flash Point 92°C (197°F)
Safety and Handling

Warning