Please login to your online account to display your discounted pricing

Ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 3, 5-dimethyl-1H-pyrrole-2-carboxylate
Name Note 98%
CAS 2199-44-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H13NO2
Formula Weight 167.21
MDL Number MFCD00010645
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC343760010

  SDS
thermo fisher scientific
343760010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC343760050

  SDS
thermo fisher scientific
AC343760050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Buff Powder
Formula Weight 167.21
Quantity 1g
Color Pale yellow to buff
Melting Point 122° to 126°C
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Ethyl 3, 5-dimethyl-1H-pyrrole-2-carboxylate
Molecular Formula C9H13NO2
MDL Number MFCD00010645
CAS 2199-44-2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.