Please login to your online account to display your discounted pricing

Ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC343760050

 View more versions of this product

Catalog No. AC343760050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 167.21
Assay ≥97.5%
Physical Form Buff Powder
Quantity 5g
Color Pale yellow to buff
Melting Point 122° to 126°C
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass Bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note 98%
CAS 2199-44-2
Chemical Name or Material Ethyl 3, 5-dimethyl-1H-pyrrole-2-carboxylate
Molecular Formula C9H13NO2
MDL Number MFCD00010645
Assay Percent Range 98%

Specifications

Chemical Name or Material Ethyl 3, 5-dimethyl-1H-pyrrole-2-carboxylate
Name Note 98%
CAS 2199-44-2
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H13NO2
Formula Weight 167.21
MDL Number MFCD00010645
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents