Please login to your online account to display your discounted pricing

Ethyl 3,5-dimethyl-1H-4-pyrazolecarboxylate, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  CC01223EA

 View more versions of this product

Catalog No. CC01223EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 168.19
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material Ethyl 3, 5-dimethyl-1H-4-pyrazolecarboxylate
Molecular Formula C8H12N2O2
CAS 35691-93-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
MDL Number MFCD00159638
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

Specifications

Chemical Name or Material Ethyl 3, 5-dimethyl-1H-4-pyrazolecarboxylate
CAS 35691-93-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H12N2O2
Formula Weight 168.19
MDL Number MFCD00159638
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

SDS & Safety Documents