missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3,5-dimethyl-1H-4-pyrazolecarboxylate, 97%, Thermo Scientific™
Chemical Identifiers
CAS
35691-93-1
Molecular Formula
C8H12N2O2
Molecular Weight (g/mol)
168.196
MDL Number
MFCD00159638
InChI Key
BCKARVLFIJPHQU-UHFFFAOYSA-N
Synonym
ethyl 3,5-dimethyl-1h-4-pyrazolecarboxylate, ethyl 3,5-dimethylpyrazole-4-carboxylate, 3,5-dimethylpyrazole-4-carboxylic acid ethyl ester, 3,5-dimethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 4-pyrazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester german, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrazole, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-, ethyl ester, dee
PubChem CID
215436
IUPAC Name
ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate
SMILES
CCOC(=O)C1=C(NN=C1C)C
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
CAS
35691-93-1
Molecular Weight (g/mol)
168.196
InChI Key
BCKARVLFIJPHQU-UHFFFAOYSA-N
PubChem CID
215436
SMILES
CCOC(=O)C1=C(NN=C1C)C
Molecular Formula
C8H12N2O2
MDL Number
MFCD00159638
Synonym
ethyl 3,5-dimethyl-1h-4-pyrazolecarboxylate, ethyl 3,5-dimethylpyrazole-4-carboxylate, 3,5-dimethylpyrazole-4-carboxylic acid ethyl ester, 3,5-dimethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 4-pyrazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester german, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrazole, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-, ethyl ester, dee
IUPAC Name
ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate
Specifications
CAS
35691-93-1
CAS Max %
100.0
Molecular Formula
C8H12N2O2
Quantity
1g
InChI Key
BCKARVLFIJPHQU-UHFFFAOYSA-N
IUPAC Name
ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate
PubChem CID
215436
Percent Purity
97%
CAS Min %
96.5
Assay Percent Range
97%
MDL Number
MFCD00159638
Synonym
ethyl 3,5-dimethyl-1h-4-pyrazolecarboxylate, ethyl 3,5-dimethylpyrazole-4-carboxylate, 3,5-dimethylpyrazole-4-carboxylic acid ethyl ester, 3,5-dimethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 4-pyrazolecarboxylic acid, 3,5-dimethyl-, ethyl ester, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester german, 3.5. dimethylpyrazol 4-carbonsaeureaethyl ester, 3,5-dimethyl-4-ethoxycarbonyl-1h-pyrazole, 1h-pyrazole-4-carboxylic acid, 3,5-dimethyl-, ethyl ester, dee
SMILES
CCOC(=O)C1=C(NN=C1C)C
Molecular Weight (g/mol)
168.196
Formula Weight
168.19
Chemical Name or Material
Ethyl 3, 5-dimethyl-1H-4-pyrazolecarboxylate
Safety and Handling
Safety and Handling
GHS H Statement: May cause respiratory irritation. Causes skin irritation. Harmful if swallowed. Causes serious eye irritation.
GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'