Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,5-dichloro-4-hydroxybenzoate, 99%

$48.07 - $482.16

Chemical Identifiers

Chemical Name or Material Ethyl 3,5-dichloro-4-hydroxybenzoate
CAS 17302-82-8
Molecular Formula C9H8Cl2O3
Formula Weight 235.07
MDL Number MFCD00016420
Beilstein 2649965
Synonym 3,5-Dichloro-4-hydroxybenzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1248906

  SDS
alfa aesar™
A1248906
5g Each for $48.07

AAA1248914

  SDS
alfa aesar™
A1248914
25g Each for $150.23

AAA1248922

  SDS
alfa aesar™
A1248922
100g Each for $482.16
Description & Specifications

Specifications

Formula Weight 235.07
Beilstein 2649965
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H8Cl2O3
RTECS Number DG7503500
Melting Point 110° to 112°C
Chemical Name or Material Ethyl 3,5-dichloro-4-hydroxybenzoate
Assay 99%
Quantity 5g
CAS 17302-82-8
Synonym 3,5-Dichloro-4-hydroxybenzoic acid ethyl ester
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00016420
Health Hazard 1 Warning

Applications
Ethyl 3,5-dichloro-4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.