Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(5-bromo-2-methoxy-3-pyridyl)acrylate

Chemical Identifiers

Chemical Name or Material Ethyl 3-(5-bromo-2-methoxy-3-pyridyl)acrylate
Molecular Formula C11H12BrNO3
Formula Weight 286.13
MDL Number MFCD07781177
Synonym 3-(5-Bromo-2-methoxy-3-pyridyl)acrylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5012003

 
alfa aesar™
H5012003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5012006

 
alfa aesar™
H5012006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 286.13
Chemical Name or Material Ethyl 3-(5-bromo-2-methoxy-3-pyridyl)acrylate
Molecular Formula C11H12BrNO3
MDL Number MFCD07781177
Synonym 3-(5-Bromo-2-methoxy-3-pyridyl)acrylic acid ethyl ester
Quantity 1g