Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,5-bis(trifluoromethyl)phenylacetate, 98%

$94.93 - $854.34

Chemical Identifiers

Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)phenylacetate
CAS 144632-97-3
Molecular Formula C12H10F6O2
Formula Weight 300.2
MDL Number MFCD08689894
Synonym 3,5-Bis(trifluoromethyl)phenylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2582003

  SDS
alfa aesar™
H2582003
1g Each for $94.93

AAH2582006

  SDS
alfa aesar™
H2582006
5g Each for $278.76

AAH2582014

  SDS
alfa aesar™
H2582014
25g Each for $854.34
Description & Specifications

Specifications

Formula Weight 300.2
MDL Number MFCD08689894
CAS 144632-97-3
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Molecular Formula C12H10F6O2
Quantity 1g
Health Hazard 1 Warning
Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)phenylacetate
Synonym 3,5-Bis(trifluoromethyl)phenylacetic acid ethyl ester
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.