Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,5-bis(trifluoromethyl)benzoate, 99%

$61.57 - $603.39

Chemical Identifiers

Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)benzoate
CAS 96617-71-9
Molecular Formula C11H8F6O2
Formula Weight 286.17
MDL Number MFCD01320684
Synonym 3,5-Bis(trifluoromethyl)benzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2395206

 
alfa aesar™
B2395206
5g Each for $163.16

AAB2395203

 
alfa aesar™
B2395203
1g Each for $61.57

AAB2395214

 
alfa aesar™
B2395214
25g Each for $603.39
Description & Specifications

Specifications

Formula Weight 286.17
CAS 96617-71-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)benzoate
Assay 99%
Molecular Formula C11H8F6O2
MDL Number MFCD01320684
Synonym 3,5-Bis(trifluoromethyl)benzoic acid ethyl ester
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.