Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(4-trimethylacetamido-3-pyridyl)acrylate

Chemical Identifiers

Chemical Name or Material Ethyl 3-(4-trimethylacetamido-3-pyridyl)acrylate
Molecular Formula C15H20N2O3
Formula Weight 276.34
MDL Number MFCD07781148
Synonym Ethyl 3-(4-pivaloylamino-3-pyridyl)acrylate; 3-(4-Trimethylacetamido-3-pyridyl)acrylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5003606

  SDS
alfa aesar™
H5003606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5003603

  SDS
alfa aesar™
H5003603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 276.34
Chemical Name or Material Ethyl 3-(4-trimethylacetamido-3-pyridyl)acrylate
Molecular Formula C15H20N2O3
MDL Number MFCD07781148
Synonym Ethyl 3-(4-pivaloylamino-3-pyridyl)acrylate; 3-(4-Trimethylacetamido-3-pyridyl)acrylic acid ethyl ester
Quantity 5g