missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(4-morpholinyl)propionate, 97%

$95.30 - $350.07

Chemical Identifiers

CAS 20120-24-5
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD00014622
InChI Key WRBIQTVRBWJCQT-UHFFFAOYSA-N
Synonym ethyl 4-morpholinepropionate, ethyl 3-morpholino propionate, ethyl 3-morpholinopropanoate, ethyl 3-4-morpholino propionate, 3-morpholin-4-yl propionic acid ethyl ester, acmc-209f5y, ethyl 3-morpholinopropionate, ethyl-3-morpholinoproprionate, ethyl3-morpholino propionate, tos-bb-1256
PubChem CID 88374
IUPAC Name ethyl 3-morpholin-4-ylpropanoate
SMILES CCOC(=O)CCN1CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0173406 View Documents Alfa Aesar™
L0173406
5g Each for $95.30
 
AAL0173414 View Documents Alfa Aesar™
L0173414
25g Each for $350.07
 
Specifications

Chemical Identifiers

20120-24-5
187.239
WRBIQTVRBWJCQT-UHFFFAOYSA-N
88374
CCOC(=O)CCN1CCOCC1
C9H17NO3
MFCD00014622
ethyl 4-morpholinepropionate, ethyl 3-morpholino propionate, ethyl 3-morpholinopropanoate, ethyl 3-4-morpholino propionate, 3-morpholin-4-yl propionic acid ethyl ester, acmc-209f5y, ethyl 3-morpholinopropionate, ethyl-3-morpholinoproprionate, ethyl3-morpholino propionate, tos-bb-1256
ethyl 3-morpholin-4-ylpropanoate

Specifications

5g
C9H17NO3
138843
WRBIQTVRBWJCQT-UHFFFAOYSA-N
ethyl 3-morpholin-4-ylpropanoate
88374
97%
20120-24-5
MFCD00014622
ethyl 4-morpholinepropionate, ethyl 3-morpholino propionate, ethyl 3-morpholinopropanoate, ethyl 3-4-morpholino propionate, 3-morpholin-4-yl propionic acid ethyl ester, acmc-209f5y, ethyl 3-morpholinopropionate, ethyl-3-morpholinoproprionate, ethyl3-morpholino propionate, tos-bb-1256
CCOC(=O)CCN1CCOCC1
187.239
187.24
Ethyl 3-(4-morpholinyl)propionate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning