Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(4-fluoroanilino)crotonate, 97%

$61.88 - $61.88

Chemical Identifiers

Chemical Name or Material Ethyl 3-(4-fluoroanilino)crotonate
CAS 18529-17-4
Molecular Formula C12H14FNO2
Formula Weight 223.25
MDL Number MFCD00016720
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1825106

  SDS
alfa aesar™
A1825106
5g Each for $61.88
Description & Specifications

Specifications

Formula Weight 223.25
Chemical Name or Material Ethyl 3-(4-fluoroanilino)crotonate
Assay 97%
Molecular Formula C12H14FNO2
MDL Number MFCD00016720
CAS 18529-17-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.