Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,4-dimethoxyphenylacetate, 98%

$109.28 - $412.53

Chemical Identifiers

Chemical Name or Material Ethyl 3,4-dimethoxyphenylacetate
CAS 18066-68-7
Molecular Formula C12H16O4
Formula Weight 224.26
MDL Number MFCD00017272
Beilstein 2697968
Synonym 3,4-Dimethoxyphenylacetic acid ethyl ester; Ethyl homoveratrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1379618

 
alfa aesar™
A1379618
50g Each for $109.28

AAA1379630

 
alfa aesar™
A1379630
250g Each for $412.53
Description & Specifications

Specifications

Formula Weight 224.26
Chemical Name or Material Ethyl 3,4-dimethoxyphenylacetate
TSCA TSCA
Assay 98%
Molecular Formula C12H16O4
MDL Number MFCD00017272
CAS 18066-68-7
Beilstein 2697968
Synonym 3,4-Dimethoxyphenylacetic acid ethyl ester; Ethyl homoveratrate
Boiling Point 159° to 160°C (4mmHg)
Quantity 50g