missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3,4-dimethoxybenzoate, 98+%, Thermo Scientific Chemicals
$102.49 - $298.27
Chemical Identifiers
CAS | 3943-77-9 |
---|---|
Molecular Formula | C11H14O4 |
Molecular Weight (g/mol) | 210.23 |
MDL Number | MFCD00015150 |
InChI Key | AYYNUGSDPRDVCH-UHFFFAOYSA-N |
Synonym | benzoic acid, 3,4-dimethoxy-, ethyl ester, ethyl veratrate, 3,4-dimethoxybenzoic acid ethyl ester, veratric acid, ethyl ester 7ci,8ci, veratric acid, ethyl ester, veratrumsaureathylester, acmc-1cme0, ethyl-3,4-dimethoxybenzoate, 4-10-00-01469 beilstein handbook reference, ksc581g5p |
PubChem CID | 77546 |
IUPAC Name | ethyl 3,4-dimethoxybenzoate |
SMILES | CCOC(=O)C1=CC=C(OC)C(OC)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0235514
|
Thermo Scientific Chemicals
L0235514 |
25 g |
Each for $102.49
|
|
|||||
AAL0235522
|
Thermo Scientific Chemicals
L0235522 |
100 g |
Each for $298.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3943-77-9 | |
210.23 | |
AYYNUGSDPRDVCH-UHFFFAOYSA-N | |
77546 | |
CCOC(=O)C1=CC=C(OC)C(OC)=C1 |
C11H14O4 | |
MFCD00015150 | |
benzoic acid, 3,4-dimethoxy-, ethyl ester, ethyl veratrate, 3,4-dimethoxybenzoic acid ethyl ester, veratric acid, ethyl ester 7ci,8ci, veratric acid, ethyl ester, veratrumsaureathylester, acmc-1cme0, ethyl-3,4-dimethoxybenzoate, 4-10-00-01469 beilstein handbook reference, ksc581g5p | |
ethyl 3,4-dimethoxybenzoate |
Specifications
3943-77-9 | |
C11H14O4 | |
25 g | |
benzoic acid, 3,4-dimethoxy-, ethyl ester, ethyl veratrate, 3,4-dimethoxybenzoic acid ethyl ester, veratric acid, ethyl ester 7ci,8ci, veratric acid, ethyl ester, veratrumsaureathylester, acmc-1cme0, ethyl-3,4-dimethoxybenzoate, 4-10-00-01469 beilstein handbook reference, ksc581g5p | |
CCOC(=O)C1=CC=C(OC)C(OC)=C1 | |
210.23 | |
210.23 | |
Ethyl 3,4-dimethoxybenzoate |
42°C to 45°C | |
MFCD00015150 | |
3118897 | |
AYYNUGSDPRDVCH-UHFFFAOYSA-N | |
ethyl 3,4-dimethoxybenzoate | |
77546 | |
≥98% |
Safety and Handling
EINECSNumber : 223-526-4
RTECSNumber : DG8690000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only