Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,4-dihydroxycinnamate, 97%

$59.38 - $724.76

Chemical Identifiers

Chemical Name or Material Ethyl 3,4-dihydroxycinnamate
CAS 66648-50-8
Molecular Formula C11H12O4
Formula Weight 208.22
MDL Number MFCD00045754
Synonym Ethyl caffeate; 3,4-Dihydroxycinnamic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2480803

  SDS
alfa aesar™
B2480803
1g Each for $59.38

AAB2480806

  SDS
alfa aesar™
B2480806
5g Each for $194.38

AAB2480814

  SDS
alfa aesar™
B2480814
25g Each for $724.76
Description & Specifications

Specifications

Formula Weight 208.22
Molecular Formula C11H12O4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Ethyl 3,4-dihydroxycinnamate
Synonym Ethyl caffeate; 3,4-Dihydroxycinnamic acid ethyl ester
Quantity 1g
MDL Number MFCD00045754
Health Hazard 1 Warning
CAS 66648-50-8

Applications
Ethyl 3,4-dihydroxycinnamate shows anti-inflammatory, immunomodulatory, and anti-carcinogenic properties. It is a potent and specific inhibitor of NF-κB and its downstream inflammatory mediators: NOS2 (inducible nitric oxide synthase; NOS; NOS II), Cox-2 (cyclooxygenase-2), and prostaglandin E2 (PGE2). Ethyl 3,4-Dihydroxycinnamate prevents DNA single-strand breaks caused by H2O2.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents Store at -20°C.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.