Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3,4-dichlorophenylacetate, 97+%

$162.73 - $162.73

Chemical Identifiers

Chemical Name or Material Ethyl 3,4-dichlorophenylacetate
CAS 6725-45-7
Molecular Formula C10H10Cl2O2
Formula Weight 233.1
MDL Number MFCD00833397
Synonym 3,4-Dichlorophenylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2156909

  SDS
alfa aesar™
B2156909
10g Each for $162.73
Description & Specifications

Specifications

Formula Weight 233.1
Chemical Name or Material Ethyl 3,4-dichlorophenylacetate
Assay ≥97%
Molecular Formula C10H10Cl2O2
MDL Number MFCD00833397
CAS 6725-45-7
Synonym 3,4-Dichlorophenylacetic acid ethyl ester
Melting Point 24° to 26°C
Quantity 10g

Applications
Ethyl 3,4-dichlorophenylacetate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.