missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 3-(4-Cyanophenyl)-3-oxopropionate, TCI America™

Manufacturer:  TCI America E10801G

 View more versions of this product

Catalog No. E10801G



Specifications

Specifications

Ethyl 3-(4-Cyanophenyl)-3-oxopropionate
Undesignated
1g
C12H11NO3
3-(4-Cyanophenyl)-3-oxopropionic Acid Ethyl Ester
CCOC(=O)CC(=O)C1=CC=C(C=C1)C#N
217.224
217.22
≥98.0% (GC,T)
140°C
67°C
49744-93-6
MFCD00029621
ITKQVWKZLCNILF-UHFFFAOYSA-N
ethyl 3-(4-cyanophenyl)-3-oxopropanoate
2757969
Crystal-Powder at 20°C

Chemical Identifiers

49744-93-6
217.224
ITKQVWKZLCNILF-UHFFFAOYSA-N
2757969
CCOC(=O)CC(=O)C1=CC=C(C=C1)C#N
C12H11NO3
MFCD00029621
3-(4-Cyanophenyl)-3-oxopropionic Acid Ethyl Ester
ethyl 3-(4-cyanophenyl)-3-oxopropanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents