missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(4-bromophenyl)propionate, 95%

Manufacturer:  Alfa Aesar™ H6163306

 View more versions of this product

Catalog No. AAH6163306



Specifications

Specifications

Ethyl 3-(4-bromophenyl)propionate
40640-98-0
MFCD07772961
STMHGPFYLQOGJD-UHFFFAOYSA-N
ethyl 3-(4-bromophenyl)propanoate
12731927
Solid
5g
C11H13BrO2
3-4-bromo-phenyl-propionic acid ethyl ester
CCOC(=O)CCC1=CC=C(C=C1)Br
257.127
257.12
95%

Chemical Identifiers

40640-98-0
257.127
STMHGPFYLQOGJD-UHFFFAOYSA-N
12731927
CCOC(=O)CCC1=CC=C(C=C1)Br
C11H13BrO2
MFCD07772961
3-4-bromo-phenyl-propionic acid ethyl ester
ethyl 3-(4-bromophenyl)propanoate
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C

Documents