missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 3-(4-bromophenyl)propionate, 95%, Thermo Scientific Chemicals
$299.89 - $990.18
Chemical Identifiers
CAS | 40640-98-0 |
---|---|
Molecular Formula | C11H13BrO2 |
Molecular Weight (g/mol) | 257.13 |
MDL Number | MFCD07772961 |
InChI Key | STMHGPFYLQOGJD-UHFFFAOYSA-N |
PubChem CID | 12731927 |
IUPAC Name | ethyl 3-(4-bromophenyl)propanoate |
SMILES | CCOC(=O)CCC1=CC=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6163306
|
Thermo Scientific Chemicals
H6163306 |
5 g |
Each for $299.89
|
|
|||||
AAH6163314
|
Thermo Scientific Chemicals
H6163314 |
25 g |
Each for $990.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40640-98-0 | |
257.13 | |
STMHGPFYLQOGJD-UHFFFAOYSA-N | |
ethyl 3-(4-bromophenyl)propanoate |
C11H13BrO2 | |
MFCD07772961 | |
12731927 | |
CCOC(=O)CCC1=CC=C(Br)C=C1 |
Specifications
40640-98-0 | |
MFCD07772961 | |
ethyl 3-4-bromophenyl propanoate, 3-4-bromophenyl propionic acid ethyl ester, benzenepropanoic acid, 4-bromo-, ethyl ester, ethyl 3-4-bromophenyl-propionate, 3-4-bromo-phenyl-propionic acid ethyl ester, ksc497s3n, ethyl3-4-bromophenyl propanoate, 4-bromobenzenepropanoic acid ethyl ester, 3-4-bromophenyl propanoic acid ethyl ester, 3-4-bromophenyl-propionic acid ethyl ester | |
CCOC(=O)CCC1=CC=C(Br)C=C1 | |
257.13 | |
257.12 | |
Solid |
C11H13BrO2 | |
5 g | |
STMHGPFYLQOGJD-UHFFFAOYSA-N | |
ethyl 3-(4-bromophenyl)propanoate | |
12731927 | |
95% | |
Ethyl 3-(4-bromophenyl)propionate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only