Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 3-(4-amino-5-bromo-3-pyridyl)acrylate, 96%

$258.59 - $258.59

Chemical Identifiers

Chemical Name or Material Ethyl 3-(4-amino-5-bromo-3-pyridyl)acrylate
CAS 912760-87-3
Molecular Formula C10H11BrN2O2
Formula Weight 271.12
MDL Number MFCD07781150
Synonym 3-(4-Amino-5-bromo-3-pyridyl)acrylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5003803

 
alfa aesar™
H5003803
1g Each for $258.59
Description & Specifications

Specifications

Formula Weight 271.12
Chemical Name or Material Ethyl 3-(4-amino-5-bromo-3-pyridyl)acrylate
Assay 96%
Molecular Formula C10H11BrN2O2
MDL Number MFCD07781150
CAS 912760-87-3
Synonym 3-(4-Amino-5-bromo-3-pyridyl)acrylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.